4'-Cyanoacetanilide 97%

Supplier: Ambeed
Warning

A204012-25G A204012-5G
77287-746EA 38 USD
77287-746 77287-748
4'-Cyanoacetanilide 97%
4'-Cyanoacetanilide

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MDL Number: MFCD00001814
CAS Number: 35704-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR