2-Bromo-1-(1,4-dimethyl-1H-pyrazol-3-yl)ethanone 97%

Supplier: Ambeed
Danger

A203991-100MG A203991-1G A203991-5G
77260-872EA 253.38 USD
77260-872 77260-874 77260-876
2-Bromo-1-(1,4-dimethyl-1H-pyrazol-3-yl)ethanone 97%
2-Bromo-1-(1,4-dimethyl-1H-pyrazol-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₉BrN₂O
MW: 217.06 g/mol
MDL Number: MFCD31619092
CAS Number: 2097938-49-1
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR