(S)-(4-Benzylmorpholin-3-yl)methanol 97%

Supplier: Ambeed
Warning

A203751-100MG A203751-1G A203751-250MG
77274-932EA 25.75 USD
77274-932 77274-934 77274-936
(S)-(4-Benzylmorpholin-3-yl)methanol 97%
(S)-(4-Benzylmorpholin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₂
MW: 207.269 g/mol
MDL Number: MFCD06799482
CAS Number: 101376-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR