(4-Fluorophenyl)hydrazine 95%

Supplier: Ambeed
Danger

A203677-5G A203677-1G A203677-25G
77287-744EA 103 USD
77287-744 77567-948 77748-288
(4-Fluorophenyl)hydrazine 95%
(4-Fluorophenyl)hydrazine

Precaución: For research use only.

Formula: C₆H₇FN₂
MW: 126.13 g/mol
MDL Number: MFCD00041262
CAS Number: 371-14-2
UN: 2922
ADR: 8,6.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR