2,4-Dihydroxy-6-methylbenzoic acid 97%

Supplier: Ambeed
Warning

A203541-1MG A203541-25MG A203541-1G
77248-886EA 5 USD
77248-886 77248-888 77593-322
2,4-Dihydroxy-6-methylbenzoic acid 97%
2,4-Dihydroxy-6-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈O₄
MDL Number: MFCD00210536
CAS Number: 480-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR