5-(4-Ethylphenyl)-1,3,4-thiadiazol-2-amine 95%

Supplier: Ambeed
Warning

A203217-100MG A203217-10G A203217-250MG A203217-25G A203217-5G
77237-208EA 9.27 USD
77237-208 77237-210 77237-212 77237-214 77237-216
5-(4-Ethylphenyl)-1,3,4-thiadiazol-2-amine 95%
5-(4-Ethylphenyl)-1,3,4-thiadiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₁N₃S
MW: 205.28 g/mol
MDL Number: MFCD02664104
CAS Number: 383130-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR