4-Fluoroindolin-2-one 97%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-2-oxindole, 4-Fluorooxindole

A203090-1G A203090-5G
77287-724EA 72.1 USD
77287-724 77287-726
4-Fluoroindolin-2-one 97%
4-Fluoroindolin-2-one

Precaución: for Research Use Only

Formula: C₈H₆FNO
MW: 151.140 g/mol
MDL Number: MFCD04112528
CAS Number: 138343-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR