1-(4,5-Dichlorothiophen-2-yl)ethanone 95%

Supplier: Ambeed
Warning

A203081-100MG A203081-1G A203081-250MG A203081-25G A203081-5G
77248-876EA 5.15 USD
77248-876 77248-878 77248-880 77248-882 77248-884
1-(4,5-Dichlorothiophen-2-yl)ethanone 95%
1-(4,5-Dichlorothiophen-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₄Cl₂OS
MW: 195.07 g/mol
MDL Number: MFCD00082793
CAS Number: 57681-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR