3',5'-Dimethoxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3,5-Dimethoxyphenyl)ethan-1-one, 1-(3,5-Dimethoxyphenyl)ethanone

A202959-100G A202959-1G A202959-250MG A202959-25G A202959-5G
77274-910EA 950.69 USD
77274-910 77274-912 77274-914 77274-916 77274-918
3',5'-Dimethoxyacetophenone 98%
3',5'-Dimethoxyacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
MDL Number: MFCD00008739
CAS Number: 39151-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR