(R)-3,3'-Bis(3,5-diphenyl)phenyl-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate ≥98%, ee 99%

Supplier: Ambeed
Warning

A202800-250MG A202800-100MG
77248-874EA 164 USD
77248-874 77593-320
(R)-3,3'-Bis(3,5-diphenyl)phenyl-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate ≥98%, ee 99%
(R)-3,3'-Bis(3,5-diphenyl)phenyl-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate

Precaución: for Research Use Only

Formula: C₅₆H₃₇O₄P
CAS Number: 361342-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR