1,2-Bis(5-methyl-2-benzoxazolyl)ethylene 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Bis(5-methylbenzo[d]oxazol-2-yl)ethene

A202499-100G A202499-25G A202499-500G A202499-5G
77248-858EA 33.99 USD
77248-858 77248-860 77248-862 77248-864
1,2-Bis(5-methyl-2-benzoxazolyl)ethylene 98%
1,2-Bis(5-methyl-2-benzoxazolyl)ethylene

Precaución: for Research Use Only

Formula: C₁₈H₁₄N₂O₂
MW: 290.32 g/mol
MDL Number: MFCD00408425
CAS Number: 1041-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR