Ethyl-3-Amino-2-methylbenzoate 98+%

Supplier: Ambeed
Warning

A202380-1G A202380-25G A202380-5G
77274-902EA 8 USD
77274-902 77274-904 77274-906
Ethyl-3-Amino-2-methylbenzoate 98+%
Ethyl-3-Amino-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00957246
CAS Number: 57414-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR