GSK-2334470 98%

Supplier: Ambeed
Warning

Synonyms: (3S,6R)-1-(6-(3-Amino-1H-indazol-6-yl)-2-(methylamino)pyrimidin-4-yl)-N-cyclohexyl-6-methylpiperidine-3-carboxamide, GSK2334470, (3S,6R)-1-[6-(3-Amino-1H-indazol-6-yl)-2-(methylamino)-4-pyrimidinyl]-N-cyclohexyl-6-methyl-3-piperidinecarboxamide

A202355-10MG A202355-1MG A202355-5MG
77237-198EA 64.89 USD
77237-198 77237-200 77237-202
GSK-2334470 98%
GSK-2334470

Precaución: for Research Use Only

Formula: C₂₅H₃₄N₈O
MW: 462.59 g/mol
MDL Number: MFCD21608525
CAS Number: 1227911-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR