trans-4-((2-Carbamoyl-5-(6,6-dimethyl-4-oxo-3-(trifluoromethyl)-4,5,6,7-tetrahydro-1H-indazol-1-yl)phenyl)amino)cyclohexyl glycinate methanesulfonate 98%

Supplier: Ambeed
Warning

A202225-10MG A202225-1MG A202225-2MG A202225-50MG A202225-5MG
77237-188EA 238.96 USD
77237-188 77237-190 77237-192 77237-194 77237-196
trans-4-((2-Carbamoyl-5-(6,6-dimethyl-4-oxo-3-(trifluoromethyl)-4,5,6,7-tetrahydro-1H-indazol-1-yl)phenyl)amino)cyclohexyl glycinate methanesulfonate 98%
trans-4-((2-Carbamoyl-5-(6,6-dimethyl-4-oxo-3-(trifluoromethyl)-4,5,6,7-tetrahydro-1H-indazol-1-yl)phenyl)amino)cyclohexyl glycinate methanesulfonate

Precaución: for Research Use Only

Formula: C₂₆H₃₄F₃N₅O₇S
MW: 617.63 g/mol
MDL Number: MFCD29924735
CAS Number: 1173111-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR