Ethyl-2,4-dihydroxy-6-methyl-3-pyridinecarboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 2,4-Dihydroxy-6-methyl-3-pyridinecarboxylate, 2,4-Dihydroxy-6-methyl-3-pyridinecarboxylic acid ethyl ester, Ethyl 2,4-dihydroxy-6-methyl-3-pyridinecarboxylate, Ethyl 2,4-Dihydroxy-6-methylnicotinate, 2,4-Dihydroxy-6-methylnicotinic acid ethyl ester
A202073-100G
A202073-10G
A202073-25G
A202073-500G
A202073-5G
77274-872EA
180.25
USD
77274-872
77274-874
77274-876
77274-878
77274-880
Ethyl-2,4-dihydroxy-6-methyl-3-pyridinecarboxylate 98%
Ethyl-2,4-dihydroxy-6-methyl-3-pyridinecarboxylate
Precaución: for Research Use Only
Formula:
C₉H₁₁NO₄ MW: 197.1900 g/mol |
MDL Number:
MFCD02929397 CAS Number: 70254-52-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...