4-((3-Chloro-2,4-difluorophenyl)amino)-7-methoxyquinazolin-6-ol 97%

Supplier: Ambeed
Warning

A201643-100MG A201643-250MG
77260-798EA 47.38 USD
77260-798 77260-800
4-((3-Chloro-2,4-difluorophenyl)amino)-7-methoxyquinazolin-6-ol 97%
4-((3-Chloro-2,4-difluorophenyl)amino)-7-methoxyquinazolin-6-ol

Precaución: for Research Use Only

Formula: C₁₅H₁₀ClF₂N₃O₂
MW: 337.71 g/mol
MDL Number: MFCD30607248
CAS Number: 1818217-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR