4-Bromoisoxazole 97%

Supplier: Ambeed
Warning

A201570-10G A201570-250MG A201570-5G A201570-1G
77287-704EA 146.26 USD
77287-704 77287-706 77287-708 77567-920
4-Bromoisoxazole 97%
4-Bromoisoxazole

Precaución: For research use only.

Formula: C₃H₂BrNO
MW: 147.9600 g/mol
MDL Number: MFCD02179569
CAS Number: 97925-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR