2-Bromo-5-methoxy-4-nitroaniline 95%

Supplier: Ambeed
Warning

A201546-1G A201546-250MG
77260-792EA 51.5 USD
77260-792 77260-794 77260-796
2-Bromo-5-methoxy-4-nitroaniline 95%
2-Bromo-5-methoxy-4-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₃
MDL Number: MFCD29038838
CAS Number: 1934530-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR