3,5-Difluoro-4-formylphenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (3,5-Difluoro-4-formylphenyl)boronic acid

A201402-10G A201402-1G A201402-250MG
77274-852EA 238.96 USD
77274-852 77274-854 77274-856
3,5-Difluoro-4-formylphenylboronic acid 95%
3,5-Difluoro-4-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₂O₃
MW: 185.9206 g/mol
MDL Number: MFCD07784344
CAS Number: 870718-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR