trans,trans-4-(4-Methylphenyl)-4'-vinylbicyclohexyl 97%

Supplier: Ambeed
Warning

Synonyms: (trans,trans)-4-(p-Tolyl)-4'-vinyl-1,1'-bi(cyclohexane), trans,trans-4-(p-Tolyl)-4'-vinylbicyclohexyl

A201246-1G A201246-5G
77237-168EA 17 USD
77237-168 77237-170
trans,trans-4-(4-Methylphenyl)-4'-vinylbicyclohexyl 97%
trans,trans-4-(4-Methylphenyl)-4'-vinylbicyclohexyl

Precaución: for Research Use Only

Formula: C₂₁H₃₀
MDL Number: MFCD12911573
CAS Number: 155041-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR