3-Fluoro-5-(trifluoromethyl)pyridine-2-carbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-5-(trifluoromethyl)picolinonitrile

A201165-100G A201165-10G A201165-1G A201165-25G A201165-5G
77274-840EA 820.91 USD
77274-840 77274-842 77274-844 77274-846 77274-848
3-Fluoro-5-(trifluoromethyl)pyridine-2-carbonitrile 98%
3-Fluoro-5-(trifluoromethyl)pyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₂F₄N₂
MW: 190.1 g/mol
MDL Number: MFCD11848039
CAS Number: 80194-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR