4-Fluoro-2-nitroaniline 98%

Supplier: Ambeed
Warning

A201155-100G A201155-10G A201155-25G A201155-500G
77287-688EA 47 USD
77287-688 77287-690 77287-692 77287-694 77287-696
4-Fluoro-2-nitroaniline 98%
4-Fluoro-2-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₅FN₂O₂
MDL Number: MFCD00007830
CAS Number: 364-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR