2-Amino-4-hydroxypyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Aminopyrimidin-4-ol

A201021-5G A201021-25G
77248-848EA 22.66 USD
77248-848 77746-254
2-Amino-4-hydroxypyrimidine 98%
2-Amino-4-hydroxypyrimidine

Precaución: for Research Use Only

Formula: C₄H₅N₃O
MW: 111.1 g/mol
MDL Number: MFCD00023256
CAS Number: 100643-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR