1-Chloroethylisopropyl carbonate 98%

Supplier: Ambeed
Danger

Synonyms: Carbonic Acid 1-Chloroethyl Isopropyl Ester, 1-Chloroethyl isopropyl carbonate

A200735-10G A200735-25G
77237-136EA 6.18 USD
77237-136 77302-714
1-Chloroethylisopropyl carbonate 98%
1-Chloroethylisopropyl carbonate

Precaución: for Research Use Only

Formula: C₆H₁₁ClO₃
MW: 166.60 g/mol
MDL Number: MFCD04038100
CAS Number: 98298-66-9
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR