3,5-Bis(trifluoromethyl)benzophenone 97%

Supplier: Ambeed
Warning

A200705-1G A200705-25G A200705-5G
77274-818EA 17.01 USD
77274-818 77274-820 77274-822
3,5-Bis(trifluoromethyl)benzophenone 97%
3,5-Bis(trifluoromethyl)benzophenone

Precaución: for Research Use Only

Formula: C₁₅H₈F₆O
MDL Number: MFCD00015510
CAS Number: 21221-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR