2-Chloro-5-methyl-1,4-phenylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-methylbenzene-1,4-diamine, 2,5-Diamino-4-chlorotoluene

A200666-25G A200666-5G
77260-788EA 17 USD
77260-788 77260-790
2-Chloro-5-methyl-1,4-phenylenediamine 98%
2-Chloro-5-methyl-1,4-phenylenediamine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂
MW: 156.61 g/mol
MDL Number: MFCD00190684
CAS Number: 5307-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR