4-Bromobutyl phenyl ether 97%

Supplier: Ambeed
Warning

A200661-100G A200661-1G A200661-25G A200661-500G A200661-5G
77287-656EA 167 USD
77287-656 77287-658 77287-660 77287-662 77287-664
4-Bromobutyl phenyl ether 97%
4-Bromobutyl phenyl ether

Precaución: for Research Use Only

Formula: (C₆H₅)O(CH₂)₄Br
MDL Number: MFCD00000262
CAS Number: 1200-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR