KM-11060 98%

Supplier: Ambeed
Warning

Synonyms: 7-Chloro-4-(4-((4-chlorophenyl)sulphonyl)piperazin-1-yl)quinoline, 7-Chloro-4-[4-[4-chlorophenyl)sulphonyl]-1-piperazinyl]quinoline, KM11060

A200589-100MG A200589-10MG A200589-1MG A200589-250MG A200589-50MG A200589-5MG
77237-124EA 277.07 USD
77237-124 77237-126 77237-128 77237-130 77237-132 77237-134
KM-11060 98%
KM-11060

Precaución: for Research Use Only

Formula: C₁₉H₁₇Cl₂N₃O₂S
MW: 422.33 g/mol
MDL Number: MFCD01524977
CAS Number: 774549-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR