N-(3-((5-chloro-2-((2-methoxy-4-(4-methylpiperazin-1-yl)phenyl)amino)pyrimidin-4-yl)oxy)phenyl)propionamide 98+%

Supplier: Ambeed
Warning

Synonyms: WZ4003

A200517-10MG A200517-25MG A200517-50MG A200517-5MG A200517-1MG A200517-100MG
77237-116EA 104.03 USD
77237-116 77237-118 77237-120 77237-122 77615-704 77746-253
N-(3-((5-chloro-2-((2-methoxy-4-(4-methylpiperazin-1-yl)phenyl)amino)pyrimidin-4-yl)oxy)phenyl)propionamide 98+%
N-(3-((5-chloro-2-((2-methoxy-4-(4-methylpiperazin-1-yl)phenyl)amino)pyrimidin-4-yl)oxy)phenyl)propionamide

Precaución: for Research Use Only

Formula: C₂₅H₂₉ClN₆O₃
MW: 496.99 g/mol
MDL Number: MFCD28015100
CAS Number: 1214265-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR