3-Fluoro-N-methoxy-N-methylbenzamide 97%

Supplier: Ambeed
Warning

A200453-1G A200453-5G
77274-800EA 22 USD
77274-800 77274-802
3-Fluoro-N-methoxy-N-methylbenzamide 97%
3-Fluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MDL Number: MFCD02684305
CAS Number: 226260-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR