9-Fluorenone-2,7-dicarboxylic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
9-Oxo-9H-fluorene-2,7-dicarboxylic acid
A200355-250MG
A200355-1G
A200355-25G
A200355-5G
A200355-100MG
77274-792EA
19.57
USD
77274-792
77593-300
77748-267
77748-268
77748-266
9-Fluorenone-2,7-dicarboxylic acid 95%
9-Fluorenone-2,7-dicarboxylic acid
Precaución: for Research Use Only
Formula:
C₁₅H₈O₅ MW: 268.23 g/mol |
MDL Number:
MFCD00016355 CAS Number: 792-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...