4,5-Dimethylthiazole 97%

Supplier: Ambeed
Danger

A200332-25G A200332-5G
77287-652EA 24 USD
77287-652 77287-654
4,5-Dimethylthiazole 97%
4,5-Dimethylthiazole

Precaución: for Research Use Only

Formula: C₅H₇NS
MDL Number: MFCD00005336
CAS Number: 3581-91-7
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR