3,4-Dimethoxypicolinic acid 97%

Supplier: Ambeed
Warning

A200286-100MG A200286-1G A200286-250MG
77260-782EA 13 USD
77260-782 77260-784 77260-786
3,4-Dimethoxypicolinic acid 97%
3,4-Dimethoxypicolinic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₄
MDL Number: MFCD18255793
CAS Number: 220437-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR