Methyl 3-(hydroxymethyl)-2-nitrobenzoate 97%

Supplier: Ambeed
Warning

A200131-100MG A200131-1G A200131-250MG
77260-776EA 71 USD
77260-776 77260-778 77260-780
Methyl 3-(hydroxymethyl)-2-nitrobenzoate 97%
Methyl 3-(hydroxymethyl)-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₉NO₅
MDL Number: MFCD30607326
CAS Number: 1331954-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR