1-(3,5-Dichloropyridin-4-yl)ethanol 97%

Supplier: Ambeed
Warning

A200126-100MG A200126-1G A200126-250MG A200126-5G
77237-102EA 31.93 USD
77237-102 77237-104 77237-106 77746-249
1-(3,5-Dichloropyridin-4-yl)ethanol 97%
1-(3,5-Dichloropyridin-4-yl)ethanol

Precaución: for Research Use Only

Formula: C₇H₇Cl₂NO
MW: 192.043 g/mol
MDL Number: MFCD23163275
CAS Number: 1254473-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR