Methyl 4-formyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 97%

Supplier: Ambeed
Warning

A199910-100MG A199910-1G A199910-250MG A199910-5G
77260-768EA 43.26 USD
77260-768 77260-770 77260-772 77260-774
Methyl 4-formyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 97%
Methyl 4-formyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₉BO₅
MW: 290.12 g/mol
MDL Number: MFCD22493774
CAS Number: 1349171-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR