2,4,6-T (2,4,6-trichlorophenoxy acetic acid) 98%

Supplier: Ambeed
Danger

A199540-10G A199540-25G A199540-5G A199540-1G
77248-826EA 68 USD
77248-826 77248-828 77248-830 77615-692
2,4,6-T (2,4,6-trichlorophenoxy acetic acid) 98%
2,4,6-T (2,4,6-trichlorophenoxy acetic acid)

Precaución: for Research Use Only

Formula: C₈H₅Cl₃O₃
MDL Number: MFCD00053498
CAS Number: 575-89-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR