2-(Aminomethyl)piperidine 95%

Supplier: Ambeed
Danger

Synonyms: 2-Piperidylmethylamine

A199530-25G A199530-5G
77287-634EA 42 USD
77287-634 77287-636
2-(Aminomethyl)piperidine 95%
2-(Aminomethyl)piperidine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
MDL Number: MFCD00129011
CAS Number: 22990-77-8
UN: 2735
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR