4-Nitro-m-toluonitrile 97%

Supplier: Ambeed
Warning

A199491-1G A199491-250MG A199491-5G A199491-25G
77237-090EA 17 USD
77237-090 77237-092 77237-094 77615-690
4-Nitro-m-toluonitrile 97%
4-Nitro-m-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MDL Number: MFCD00017615
CAS Number: 96784-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR