(3,4-Dichlorophenoxy)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3,4-D, 2-(3,4-Dichlorophenoxy)acetic acid

A199112-5G A199112-10G A199112-1G A199112-250MG
77274-766EA 59.74 USD
77274-766 77748-260 77748-261 77748-262
(3,4-Dichlorophenoxy)acetic acid 97%
(3,4-Dichlorophenoxy)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃Cl₂)OCH₂COOH
MW: 221.04 g/mol
MDL Number: MFCD00004303
CAS Number: 588-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR