3-Amino-1,1,1-trifluoropropan-2-ol 96%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-3,3,3-trifluoropropylamine, 3-Amino-2-hydroxy-1,1,1-trifluoropropane

A198968-1G A198968-250MG A198968-25G A198968-5G
77274-758EA 46.35 USD
77274-758 77274-760 77274-762 77274-764
3-Amino-1,1,1-trifluoropropan-2-ol 96%
3-Amino-1,1,1-trifluoropropan-2-ol

Precaución: for Research Use Only

Formula: C₃H₆F₃NO
MW: 129.081 g/mol
MDL Number: MFCD02082679
CAS Number: 431-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR