3-Nitro-p-toluonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-3-nitrobenzonitrile

A198919-100G A198919-10G A198919-25G A198919-500G A198919-5G
77274-748EA 80.34 USD
77274-748 77274-750 77274-752 77274-754 77274-756
3-Nitro-p-toluonitrile 97%
3-Nitro-p-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD00031482
CAS Number: 939-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR