2-Acetamido-4-phenylbutanoic acid 97%

Supplier: Ambeed
Warning

A198819-1G A198819-250MG
77248-816EA 23 USD
77248-816 77248-818
2-Acetamido-4-phenylbutanoic acid 97%
2-Acetamido-4-phenylbutanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD18862874
CAS Number: 60505-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR