3-Fluoro-5-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-5-methoxybenzeneboronic acid

A198788-100G A198788-100MG A198788-10G A198788-1G A198788-250MG A198788-25G A198788-5G
77274-734EA 267 USD
77274-734 77274-736 77274-738 77274-740 77274-742 77274-744 77274-746
3-Fluoro-5-methoxyphenylboronic acid 98%
3-Fluoro-5-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₃
MW: 169.95 g/mol
MDL Number: MFCD07363780
CAS Number: 609807-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR