N-(2-(4-Chloro-2-methylphenoxy)ethyl)thiophene-2-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: ML402

A198458-10MG A198458-1MG A198458-25MG A198458-5MG
77274-724EA 17.51 USD
77274-724 77274-726 77274-728 77274-730
N-(2-(4-Chloro-2-methylphenoxy)ethyl)thiophene-2-carboxamide 98%
N-(2-(4-Chloro-2-methylphenoxy)ethyl)thiophene-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₄H₁₄ClNO₂S
MW: 295.78 g/mol
MDL Number: MFCD01990273
CAS Number: 298684-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR