2-Amino-6-fluorobenzothiazole 96%

Supplier: Ambeed
Warning

Synonyms: 6-Fluorobenzothiazol-2-ylamine, 6-Fluorobenzo[d]thiazol-2-amine, 6-Fluoro-1,3-benzothiazol-2-amine

A198110-100G A198110-10G A198110-25G A198110-5G
77274-706EA 141.11 USD
77274-706 77274-708 77274-710 77274-712
2-Amino-6-fluorobenzothiazole 96%
2-Amino-6-fluorobenzothiazole

Precaución: for Research Use Only

Formula: C₇H₅FN₂S
MW: 168.19 g/mol
MDL Number: MFCD00013336
CAS Number: 348-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR