Methyl-2-(piperazin-1-yl)benzoate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-(piperazin-1-yl)benzoate
A198030-100MG
A198030-1G
A198030-250MG
77274-700EA
31.93
USD
77274-700
77274-702
77274-704
Methyl-2-(piperazin-1-yl)benzoate 95%
Methyl-2-(piperazin-1-yl)benzoate
Precaución: for Research Use Only
Formula:
C₁₂H₁₆N₂O₂ MW: 220.2700 g/mol |
MDL Number:
MFCD06656931 CAS Number: 159974-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...