4-Bromo-3,5-dimethoxybenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (4-Bromo-3,5-dimethoxyphenyl)methanol

A197834-1G A197834-250MG A197834-5G
77274-694EA 8.24 USD
77274-694 77274-696 77274-698
4-Bromo-3,5-dimethoxybenzyl alcohol 98%
4-Bromo-3,5-dimethoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₃
MW: 247.086 g/mol
MDL Number: MFCD06658150
CAS Number: 61367-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR