3-Cyano-4-methyl-2-pyridone 98%

Supplier: Ambeed
Warning

A197810-1G A197810-250MG A197810-25G A197810-5G
77260-718EA 52.53 USD
77260-718 77260-720 77260-722 77260-724
3-Cyano-4-methyl-2-pyridone 98%
3-Cyano-4-methyl-2-pyridone

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.13 g/mol
MDL Number: MFCD00716620
CAS Number: 93271-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR