2-(3-Methyl-1H-1,2,4-triazol-5-yl)pyridine 97%

Supplier: Ambeed
Warning

A197808-250MG A197808-5G
77260-714EA 21.63 USD
77260-714 77260-716
2-(3-Methyl-1H-1,2,4-triazol-5-yl)pyridine 97%
2-(3-Methyl-1H-1,2,4-triazol-5-yl)pyridine

Precaución: for Research Use Only

Formula: C₈H₈N₄
MW: 160.18 g/mol
MDL Number: MFCD11106659
CAS Number: 25433-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR